Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163290.1 | NP_001156762.1 | 11 | 1323 | ||
NM_023341.3 | NP_075830.2 | 11 | 1421 | |||
GenBank mRNA | AJ278735.1 | - | 11 | 1301 | ||
AK004177.1 | - | 4 | 306 | |||
AK004791.1 | - | 11 | 1363 | |||
AK014605.2 | - | 11 | 1363 | |||
AK191635.1 | - | 2 | 46 | |||
AK192575.1 | - | 2 | 46 | |||
AK194890.1 | - | 2 | 46 | |||
AK196005.1 | - | 2 | 46 | |||
AK199890.1 | - | 46 | ||||
AK200253.1 | - | 2 | 46 | |||
AK202430.1 | - | 2 | 46 | |||
AK207786.1 | - | 2 | 46 | |||
AK217974.1 | - | 3 | 161 | |||
BC030937.1 | - | 11 | 1369 | 4922892 | ||
U31629.1 | - | 9 | 962 |