Search
Search

Ttc33| Gene Symbol: Ttc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026213.3 | NP_080489.1 | 5 | 752 | ||
| GenBank mRNA | AK013464.1 | - | 5 | 598 | ||
| AK050426.1 | - | 5 | 592 | |||
| AK075643.1 | - | 5 | 578 | |||
| BC017533.1 | - | 4 | 517 | |||
| BC061018.1 | - | 5 | 752 | 6590550 | ||