Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026553.4 | NP_080829.1 | 6 | 717 | ||
GenBank mRNA | AK002259.1 | - | 2 | 122 | ||
AK011071.1 | - | 6 | 712 | |||
AK077329.1 | - | 6 | 703 | |||
AK160409.1 | - | 6 | 670 | |||
BC011117.1 | - | 6 | 659 | 4190535 |