This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
Ak8
- Gene Name:
-
adenylate kinase 8
- Gene Aliases:
-
1190002A17Rik, AK 8
- Chromosome Location:
-
Chr.2: 28698226 - 28813165 on Build GRCm38
- UniGene ID:
-
Mm.273318
- Species:
- Mouse
- Species Specific ID (Flybase ID):
-
-
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System