Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Comtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026965.2 | NP_081241.1 | 3, 4 | 355 | ||
GenBank mRNA | AK007659.1 | - | 3, 4 | 353 | ||
AK054334.1 | - | 3, 4 | 356 | |||
BC019467.1 | - | 3, 4 | 339 | |||
BC049670.1 | - | 3, 4 | 350 | 6535487 | ||
CT010388.1 | - | 3, 4 | 317 |
Gene Symbol: A430057M04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046931.1 | - | 641 |