Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145692.2 | NP_663730.1 | 2 | 173 | ||
GenBank mRNA | AK005871.1 | - | 2 | 168 | ||
AK015164.1 | - | 2 | 168 | |||
AK076637.1 | - | 2 | 174 | |||
BC022722.1 | - | 2 | 157 | 4503172 |