siRNA Details
Gene Symbol: 4930542D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016020.1 | - | 1 | 154 |
Gene Symbol: Ccdc54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027046.3 | NP_081322.1 | 1 | 503 | ||
GenBank mRNA | AK005741.1 | - | 1 | 439 | ||
BC049741.1 | - | 1 | 503 | 6771599 |
