Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027268.2 | NP_081544.1 | 5 | 757 | ||
GenBank mRNA | AK012765.1 | - | 5 | 785 | ||
AK032573.1 | - | 5 | 784 | |||
AK050481.1 | - | 5 | 775 | |||
AK082953.1 | - | 5 | 757 | |||
AK089968.1 | - | 5 | 747 | |||
AK129084.1 | - | 5 | 744 | |||
AK145904.1 | - | 5 | 759 | |||
BC023889.1 | - | 5 | 744 | 5252046 | ||
EF410099.1 | - | 708 |