Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039189.2 | NP_001034278.1 | 4 | 799 | ||
NM_024274.3 | NP_077236.1 | 5 | 1432 | |||
GenBank mRNA | AK013211.1 | - | 5 | 1171 | ||
AK020164.1 | - | 4 | 585 | |||
AK054370.1 | - | 1198 | ||||
AK076259.1 | - | 5 | 1217 | |||
AK170986.1 | - | 5 | 1215 | |||
BC002147.1 | - | 5 | 1202 | 3484842 |