Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OPTN SIRNA Assays ›
Gene Symbol:
Optn
Gene Name:
optineurin
Gene Aliases:
4930441O07Rik, FIP2, HYPL, NRP
Chromosome Location:
Chr.2: 5020520 - 5064112 on Build GRCm38
UniGene ID:
Mm.116942
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS230851
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Optn
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181848.4
NP_862896.1
3
222
GenBank mRNA
AY071834.1
-
3
329
BC061185.1
-
3
222
30314331
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_145081, XM_006254263, XM_006254262, XM_006254264, XM_008771853, and others.
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to Golgi
negative regulation of receptor recycling
autophagy
Golgi organization
positive regulation of cell proliferation
macroautophagy
cellular protein localization
Golgi to plasma membrane protein transport
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of neuron apoptotic process
cytokine secretion
defense response to Gram-negative bacterium
protein homooligomerization
Golgi ribbon formation
TFIIIA-class transcription factor binding
protein binding
protein C-terminus binding
Rab GTPase binding
polyubiquitin binding
identical protein binding
ubiquitin binding
K63-linked polyubiquitin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0