Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdsub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159636.1 | NP_001153108.1 | 9 | 1202 | ||
NM_028118.2 | NP_082394.1 | 9 | 1202 | |||
GenBank mRNA | AK011391.1 | - | 9 | 1138 | ||
BC039804.1 | - | 2 | 58 | |||
BC050792.1 | - | 9 | 1143 | |||
BC061948.1 | - | 9 | 1180 | 6312489 |