Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028480.2 | NP_082756.1 | 3, 4 | 973 | ||
GenBank mRNA | AK005559.1 | - | 3, 4 | 979 | ||
AK145976.1 | - | 3, 4 | 974 | |||
AK146102.1 | - | 3, 4 | 974 | |||
BC100349.1 | - | 3, 4 | 950 |