Search
Search

Cmtm2a| Gene Symbol: Cmtm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027022.4 | NP_081298.1 | 3 | 496 | ||
| GenBank mRNA | AK005603.1 | - | 3 | 449 | ||
| AK161303.1 | - | 3 | 490 | |||
| AY172740.1 | - | 3 | 377 | |||
| AY241871.1 | - | 3 | 496 | |||
| BC049747.1 | - | 3 | 489 | 6772661 | ||