Search
Search

Cmtm2a| Gene Symbol: Cmtm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027022.4 | NP_081298.1 | 1 | 138 | ||
| GenBank mRNA | AK005603.1 | - | 1 | 91 | ||
| AK006874.1 | - | 1 | 139 | |||
| AK161303.1 | - | 1 | 132 | |||
| AY162281.1 | - | 1 | 19 | |||
| AY172740.1 | - | 1 | 19 | |||
| AY241871.1 | - | 1 | 138 | |||
| BC049747.1 | - | 1 | 131 | 6772661 | ||