Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153198.2 | NP_694878.2 | 9 | 1519 | ||
NM_177993.3 | NP_818774.2 | 9 | 1519 | |||
GenBank mRNA | AK028674.1 | - | 9 | 1207 | ||
AK028949.1 | - | 9 | 1205 | |||
AK082770.1 | - | 7 | 913 | |||
AK132503.1 | - | 9 | 1206 | |||
AK133127.1 | - | 9 | 1520 | |||
AK139211.1 | - | 9 | 1205 | |||
AK142187.1 | - | 9 | 1205 | |||
BC026853.1 | - | 9 | 1207 | 4458743 |