Search
Search

Zfp763| Gene Symbol: Zfp763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028543.3 | NP_082819.1 | 3, 4 | 287 | ||
| GenBank mRNA | AK006897.1 | - | 3, 4 | 339 | ||
| AK088750.1 | - | 3, 4 | 288 | |||
| AK141711.1 | - | 3, 4 | 288 | |||
| BC056193.1 | - | 3, 4 | 274 | 30011543 | ||