Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079814.1 | NP_001073282.1 | 10 | 900 | ||
NM_028720.2 | NP_082996.2 | 10 | 900 | |||
GenBank mRNA | AK014456.1 | - | 10 | 840 | ||
AK040841.1 | - | 10 | 850 | |||
AK140812.1 | - | 8 | 2498 | |||
AK145277.1 | - | 10 | 851 | |||
AK150349.1 | - | 10 | 844 | |||
AK151832.1 | - | 10 | 847 | |||
AK152586.1 | - | 10 | 847 | |||
AK152887.1 | - | 10 | 844 | |||
AK159476.1 | - | 10 | 854 | |||
AK188170.1 | - | 5 | 287 | |||
AK189966.1 | - | 6 | 333 | |||
AK208357.1 | - | 6 | 331 | |||
BC006893.1 | - | 10 | 834 | 3599477 | ||
CF174076.1 | - | 5 | 238 |