Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028900.4 | NP_083176.3 | 1 | 1321 | ||
GenBank mRNA | AK016531.1 | - | 1270 | |||
AK032253.1 | - | 1 | 1321 | |||
AK181400.1 | - | 1 | 232 | |||
BC066807.1 | - | 1 | 1290 | 30101518 |