Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133858.4 | NP_598619.2 | 9 | 2261 | ||
NM_199475.1 | NP_955769.1 | 9 | 2288 | |||
GenBank mRNA | AB075603.1 | - | 8 | 1299 | ||
AK036838.1 | - | 9 | 2253 | |||
AK087859.1 | - | 9 | 2104 | |||
AK169673.1 | - | 9 | 2288 | |||
AK177378.1 | - | 1 | 72 | |||
BC008112.1 | - | 2 | 202 | |||
BC039762.1 | - | 8 | 1106 | |||
BC051048.1 | - | 8 | 1171 | |||
BC057901.1 | - | 7 | 790 | |||
BC069953.1 | - | 2 | 292 | |||
BC093521.1 | - | 5 | 857 | 4007022 | ||
BC094388.1 | - | 9 | 2105 | 5708548 | ||
BC132301.1 | - | 8 | 1533 | 40130578 | ||
BC144749.1 | - | 8 | 1533 | 9053297 |