Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146094.2 | NP_666206.1 | 9 | 1214 | ||
GenBank mRNA | AB072976.1 | - | 9 | 1129 | ||
AF061504.1 | - | 9 | 1116 | |||
AK033308.1 | - | 7 | 571 | |||
AK083959.1 | - | 9 | 1076 | |||
AK154367.1 | - | 9 | 1164 | |||
BC022139.1 | - | 9 | 1159 | 5100199 | ||
BC026831.1 | - | 9 | 1181 | 4239695 | ||
BC026848.1 | - | 9 | 1154 | 4196030 | ||
BC063053.1 | - | 9 | 1074 | 5696722 | ||
KF987079.1 | - | 9 | 1098 |