siRNA Details
Gene Symbol: Ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029665.3 | NP_083941.2 | 11 | 1218 | ||
GenBank mRNA | AK044487.1 | - | 11 | 1250 | ||
BC029746.1 | - | 11 | 1218 | 3494321 | ||
BC031900.1 | - | 11 | 1154 | 5364208 |
