Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029749.2 | NP_084025.2 | 11 | 1312 | ||
GenBank mRNA | AK010801.1 | - | 10 | 1122 | ||
AY336567.1 | - | 10 | 1185 | |||
BC137852.1 | - | 10 | 1205 | 8860871 | ||
BM940384.1 | - | 3 | 227 | |||
CF532651.1 | - | 2 | 211 |