Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252459.1 | NP_001239388.1 | 22 | 2866 | ||
NM_001252460.1 | NP_001239389.1 | 22 | 2921 | |||
NM_133769.3 | NP_598530.2 | 22 | 2629 | |||
GenBank mRNA | AB093288.2 | - | 21 | 2501 | ||
AF334144.1 | - | 22 | 2564 | |||
AK030397.1 | - | 22 | 2566 | |||
AK147224.1 | - | 22 | 2867 | |||
AK147586.1 | - | 22 | 2622 | |||
AK147632.1 | - | 22 | 2563 | |||
AK147971.1 | - | 22 | 2569 | |||
AK197410.1 | - | 3 | 303 | |||
AK211920.1 | - | 2 | 122 | |||
BC038080.1 | - | 5 | 441 | |||
BC056974.1 | - | 22 | 2577 | 6831770 |