Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029850.3 | NP_084126.1 | 3, 4 | 446 | ||
GenBank mRNA | AK014498.1 | - | 3, 4 | 389 | ||
AK044782.1 | - | 3, 4 | 417 | |||
AK086803.1 | - | 3, 4 | 443 | |||
BC038481.1 | - | 3, 4 | 371 | 4501205 | ||
BC046777.1 | - | 3, 4 | 276 | 5709511 |