Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029891.2 | NP_084167.2 | 3 | 2092 | ||
GenBank mRNA | AK079123.1 | - | 1 | 873 | ||
BC095972.1 | - | 1 | 851 | |||
BC138828.1 | - | 3 | 2060 | 8861850 | ||
BC138829.1 | - | 3 | 2060 | 8861851 |