Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198163.1 | NP_937806.1 | 4 | 463 | ||
GenBank mRNA | AB232631.1 | - | 4 | 303 | ||
AK156576.1 | - | 4 | 454 | |||
AK213175.1 | - | 1 | 77 | |||
BC030941.1 | - | 2 | 154 | |||
BC056466.1 | - | 4 | 463 | 5717246 |