Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029920.7 | NP_084196.4 | 12, 13 | 4568 | ||
GenBank mRNA | AB093263.1 | - | 11, 12 | 3442 | ||
AK021162.1 | - | 6, 7 | 770 | |||
AK147457.1 | - | 12, 13 | 4569 | |||
BC055016.1 | - | 6, 7 | 811 | |||
BC085194.1 | - | 12, 13 | 3914 |