Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081222.1 | NP_001074691.1 | 7 | 2934 | ||
GenBank mRNA | AK040576.1 | - | 7 | 762 | ||
AK049589.1 | - | 6 | 650 | |||
AK139549.1 | - | 7 | 2658 | |||
AK145333.1 | - | 7 | 2935 | |||
AK145870.1 | - | 6 | 582 | |||
AK173297.1 | - | 5 | 2314 | |||
BC008220.1 | - | 5 | 684 | |||
BC076613.1 | - | 7 | 2664 | |||
BC151069.1 | - | 7 | 2890 | 9087982 | ||
BC151077.1 | - | 7 | 2890 | 9087990 | ||
BC172153.1 | - | 7 | 2890 |