Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190406.1 | NP_001177335.1 | 2 | 689 | ||
NM_001190408.1 | NP_001177337.1 | 2 | 689 | |||
NM_030228.3 | NP_084504.2 | 3 | 947 | |||
NM_144560.3 | NP_653146.1 | 3 | 947 | |||
GenBank mRNA | AF508323.1 | - | 1 | 591 | ||
AF508324.1 | - | 1 | 591 | |||
AK019661.1 | - | 3 | 922 | |||
AK052229.1 | - | 3 | 948 | |||
AK082864.1 | - | 2 | 657 | |||
BC031785.1 | - | 2 | 639 | 4194333 |