Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085495.2 | NP_001078964.1 | 39 | 5445 | ||
GenBank mRNA | AK029778.1 | - | 16 | 2070 | ||
AK084239.1 | - | 16 | 2084 | |||
AK155320.1 | - | 24 | 3086 | |||
BC158012.1 | - | 39 | 5440 | 9088150 |