Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037754.3 | NP_001032843.1 | 7 | 725 | ||
NM_001037755.3 | NP_001032844.2 | 7 | 725 | |||
NM_130862.4 | NP_570932.2 | 7 | 725 | |||
GenBank mRNA | AF390178.1 | - | 7 | 683 | ||
AF390179.1 | - | 7 | 683 | |||
AK049469.1 | - | 7 | 726 | |||
AK145924.1 | - | 7 | 726 | |||
AK158579.1 | - | 7 | 750 | |||
AK160401.1 | - | 7 | 726 | |||
AK205197.1 | - | 1 | 89 | |||
BC006620.1 | - | 7 | 716 | 3158004 | ||
BC016411.1 | - | 7 | 716 | 3158004 |