Search
Search

Galnt2| Gene Symbol: Galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139272.2 | NP_644678.2 | 5 | 630 | ||
| GenBank mRNA | AF348968.1 | - | 5 | 498 | ||
| AK046567.1 | - | 5 | 588 | |||
| AK145122.1 | - | 5 | 508 | |||
| AK149609.1 | - | 5 | 556 | |||
| AK151536.1 | - | 5 | 421 | |||
| BC007172.1 | - | 4 | 365 | |||
| BC053063.1 | - | 4 | 473 | 5702572 | ||
| BC059818.1 | - | 5 | 575 | 5697789 | ||