Search
Search

Abl1| Gene Symbol: Abl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112703.2 | NP_001106174.1 | 6 | 2290 | ||
| NM_001283045.1 | NP_001269974.1 | 6 | 1167 | |||
| NM_001283046.1 | NP_001269975.1 | 7 | 2495 | |||
| NM_001283047.1 | NP_001269976.1 | 7 | 2434 | |||
| NM_009594.4 | NP_033724.2 | 6 | 1061 | |||
| GenBank mRNA | AK088252.1 | - | 7 | 2420 | ||
| AK090095.1 | - | 6 | 1019 | |||
| AK166028.1 | - | 6 | 2279 | |||
| AK166210.1 | - | 7 | 2529 | |||
| BC059260.1 | - | 6 | 1088 | 6408990 | ||
| BC103770.1 | - | 6 | 1514 | 30848553 | ||
| CF536749.1 | - | 4 | 664 | |||
| CF727094.1 | - | 3 | 194 | |||
| CK626997.1 | - | 3 | 213 | |||
| J02995.1 | - | 6 | 1014 | |||
| L10656.1 | - | 4 | 615 | |||