Search
Search

Cfd| Gene Symbol: Cfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291915.1 | NP_001278844.1 | 3 | 352 | ||
| NM_013459.3 | NP_038487.1 | 3 | 355 | |||
| GenBank mRNA | AK143817.1 | - | 3 | 299 | ||
| BC138778.1 | - | 3 | 355 | 8861800 | ||
| BC138779.1 | - | 3 | 355 | 8861801 | ||
| BC145412.1 | - | 3 | 352 | 9053970 | ||
| DV051476.1 | - | 3 | 300 | |||
| M11768.1 | - | 3 | 296 | |||