Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110505.1 | NP_001103975.1 | 3 | 323 | ||
NM_007446.2 | NP_031472.2 | 3 | 432 | |||
GenBank mRNA | AK029642.1 | - | 3 | 355 | ||
BC009121.1 | - | 3 | 306 | 3710941 | ||
CK791844.1 | - | 3 | 365 | |||
J00356.1 | - | 2 | 276 | |||
V00717.1 | - | 3 | 321 | |||
V00719.1 | - | 3 | 432 | |||
V00720.1 | - | 3 | 323 |