Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031259.1 | - | 4 | 389 | ||
AK042977.1 | - | 4 | 429 | |||
AK053123.1 | - | 4 | 847 | |||
AK077591.1 | - | 4 | 402 | |||
AK167133.1 | - | 4 | 399 | |||
AK167601.1 | - | 4 | 403 | |||
AK167653.1 | - | 4 | 402 | |||
AK169094.1 | - | 4 | 399 | |||
AK169412.1 | - | 4 | 399 | |||
BC010487.1 | - | 4 | 682 | 3483748 | ||
D87899.1 | - | 4 | 394 |