Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art2a-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007490.1 | NP_031516.1 | 497 | |||
GenBank mRNA | AF016462.1 | - | 497 | |||
BC120753.1 | - | 591 | 40129676 | |||
X52991.1 | - | 497 |