Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007555.3 | NP_031581.2 | 3 | 1462 | ||
GenBank mRNA | AK033362.1 | - | 3 | 1465 | ||
AK160412.1 | - | 570 | ||||
BC100751.1 | - | 3 | 727 | |||
BC100752.1 | - | 3 | 727 | |||
BC100753.1 | - | 3 | 733 | |||
BC100754.1 | - | 3 | 727 | |||
BC141283.1 | - | 3 | 1259 | 9055895 | ||
L41145.1 | - | 3 | 728 |