Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009759.4 | NP_033889.2 | 17 | 1834 | ||
GenBank mRNA | AF012104.1 | - | 16 | 1687 | ||
AK036707.1 | - | 16 | 1693 | |||
AK040936.1 | - | 17 | 1835 | |||
BC138104.1 | - | 17 | 1711 | 8861123 | ||
BC138105.1 | - | 17 | 1711 | 8861124 | ||
U88091.1 | - | 17 | 1820 |