Search
Search

Cd3d| Gene Symbol: Cd3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013487.3 | NP_038515.3 | 1 | 124 | ||
| GenBank mRNA | AK042172.1 | - | 1 | 126 | ||
| AK050629.1 | - | 1 | 113 | |||
| AK153843.1 | - | 1 | 89 | |||
| AK154064.1 | - | 1 | 86 | |||
| BC062807.1 | - | 1 | 29 | 1366111 | ||
| EF382667.1 | - | 1 | 24 | |||
| HM536173.1 | - | 1 | 24 | |||