Search
Search

Slc29a2| Gene Symbol: Slc29a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007854.3 | NP_031880.2 | 6 | 855 | ||
| GenBank mRNA | AF183397.1 | - | 6 | 765 | ||
| AK010950.1 | - | 6 | 792 | |||
| AK083484.1 | - | 6 | 752 | |||
| AK144451.1 | - | 6 | 748 | |||
| AK154310.1 | - | 6 | 790 | |||
| BC048958.1 | - | 6 | 1001 | 6491511 | ||
| X86682.1 | - | 4 | 423 | |||