Search
Search

Emb| Gene Symbol: Emb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010330.4 | NP_034460.3 | 6 | 1092 | ||
| GenBank mRNA | AK079570.1 | - | 6 | 1034 | ||
| AK088480.1 | - | 6 | 920 | |||
| AK138271.1 | - | 6 | 1041 | |||
| AK148449.1 | - | 6 | 920 | |||
| AK148540.1 | - | 6 | 920 | |||
| AK150625.1 | - | 6 | 921 | |||
| AK161679.1 | - | 6 | 920 | |||
| AK166602.1 | - | 6 | 920 | |||
| AK167997.1 | - | 6 | 952 | |||
| BC014858.1 | - | 6 | 911 | 4500922 | ||
| J03535.2 | - | 6 | 919 | |||