Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLOT1 SIRNA Assays ›
          Gene Symbol:
          Flot1
          Gene Name:
          flotillin 1
          Gene Aliases:
          reggie-2
          Chromosome Location:
          Chr.17: 35823320 - 35832787 on Build GRCm38
          UniGene ID:
          Mm.2931
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS236529
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Flot1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008027.2 NP_032053.1 6, 7 626
          GenBank mRNA AF145044.1 - 5, 6 467
          AK150692.1 - 6, 7 648
          AK150827.1 - 6, 7 575
          AK167323.1 - 6, 7 553
          AK181092.1 - 3, 4 296
          AY167925.1 - 6, 7 632
          BC004647.1 - 6, 7 636 3158071
          U90435.1 - 6, 7 626

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          membrane raft assembly
          positive regulation of cytokine production
          positive regulation of protein phosphorylation
          regulation of receptor internalization
          endocytosis
          axonogenesis
          axon guidance
          extracellular matrix disassembly
          positive regulation of protein binding
          positive regulation of interferon-beta production
          dsRNA transport
          positive regulation of heterotypic cell-cell adhesion
          positive regulation of toll-like receptor 3 signaling pathway
          response to endoplasmic reticulum stress
          regulation of Rho protein signal transduction
          plasma membrane raft assembly
          positive regulation of endocytosis
          positive regulation of skeletal muscle tissue development
          protein homooligomerization
          positive regulation of cell adhesion molecule production
          protein kinase C signaling
          cellular response to exogenous dsRNA
          protein localization to plasma membrane
          establishment of protein localization to plasma membrane
          positive regulation of myoblast fusion
          positive regulation of cell junction assembly
          protein localization to membrane raft
          positive regulation of cell-cell adhesion mediated by cadherin
          protease binding
          protein binding
          ionotropic glutamate receptor binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0