Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008189.2 | NP_032215.2 | 1 | 279 | ||
GenBank mRNA | BC026834.1 | - | 1 | 279 | 4503333 | |
BC031810.1 | - | 1 | 246 | 4501021 | ||
L36860.1 | - | 1 | 136 |