Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008491.1 | NP_032517.1 | 4 | 437 | ||
GenBank mRNA | AK002932.1 | - | 4 | 470 | ||
AK149774.1 | - | 4 | 469 | |||
BC020275.1 | - | 4 | 437 | |||
BC132069.1 | - | 4 | 437 | 40130346 | ||
BC132071.1 | - | 4 | 437 | 40130348 | ||
S82469.1 | - | 4 | 401 | |||
X14607.1 | - | 4 | 437 |