Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111096.1 | NP_001104566.1 | 13 | 1750 | ||
NM_010747.2 | NP_034877.2 | 13 | 1687 | |||
GenBank mRNA | AK028112.1 | - | 13 | 1683 | ||
AK151914.1 | - | 13 | 1746 | |||
AK152199.1 | - | 13 | 1748 | |||
AK153038.1 | - | 13 | 1684 | |||
AK170561.1 | - | 13 | 1748 | |||
BC003253.1 | - | 13 | 1627 | 3500419 | ||
BC031547.1 | - | 13 | 1720 | 3589207 | ||
BC157998.1 | - | 13 | 1519 | 9007404 | ||
M57696.1 | - | 13 | 1586 | |||
M57697.1 | - | 13 | 1523 | |||
M64608.1 | - | 13 | 1736 |