Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Men1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168488.1 | NP_001161960.1 | 2 | 246 | ||
NM_001168489.1 | NP_001161961.1 | 2 | 224 | |||
NM_001168490.1 | NP_001161962.1 | 1 | 560 | |||
NM_008583.2 | NP_032609.1 | 1 | 560 | |||
GenBank mRNA | AB023401.1 | - | 1 | 187 | ||
AF016398.1 | - | 2 | 199 | |||
AF072755.1 | - | 2 | 183 | |||
AF109389.1 | - | 1 | 559 | |||
AF130368.1 | - | 1 | 542 | |||
AK133984.1 | - | 2 | 182 | |||
AK154371.1 | - | 2 | 205 | |||
AK164700.1 | - | 2 | 181 | |||
AK170713.1 | - | 1 | 540 | |||
BC006610.1 | - | 2 | 179 | |||
BC036287.1 | - | 1 | 488 | 4189611 | ||
CF170561.1 | - | 2 | 202 | |||
CX568988.1 | - | 1 | 232 |