Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CITED2 SIRNA Assays ›
          Gene Symbol:
          Cited2
          Gene Name:
          Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2
          Gene Aliases:
          AI835299, ER154-like, Mrg1, Msg2, p35srj
          Chromosome Location:
          Chr.10: 17723205 - 17725674 on Build GRCm38
          UniGene ID:
          Mm.272321
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS237298
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cited2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010828.3 NP_034958.2 2 252
          GenBank mRNA AK131664.1 - 2 255
          AK133791.1 - 2 253
          AK183582.1 - 2 59
          AK187598.1 - 2 182
          BC057126.1 - 2 234 6415181
          U86445.1 - 2 232
          Y15163.1 - 2 231

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001168389, NM_006079, NM_001168388, AF109161, U65093, and others.
          This siRNA also targets the following transcripts in Rat: NM_053698, AF361476, BC087005.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          vasculogenesis
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          kidney development
          trophectodermal cell differentiation
          neural tube formation
          neural tube closure
          liver development
          embryonic placenta development
          vasculature development
          heart looping
          morphogenesis of an epithelium
          lens morphogenesis in camera-type eye
          hematopoietic progenitor cell differentiation
          leukocyte differentiation
          outflow tract morphogenesis
          regulation of organ formation
          endocardial cushion development
          ventricular septum development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          determination of left/right symmetry
          central nervous system development
          peripheral nervous system development
          heart development
          sex determination
          cell proliferation
          male gonad development
          positive regulation of gene expression
          negative regulation of gene expression
          cranial nerve morphogenesis
          positive regulation of cell-cell adhesion
          cell differentiation
          adrenal gland development
          negative regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          granulocyte differentiation
          response to fluid shear stress
          positive regulation of peroxisome proliferator activated receptor signaling pathway
          adrenal cortex formation
          skeletal muscle cell differentiation
          negative regulation of apoptotic process
          response to estrogen
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          spleen development
          thymus development
          embryonic camera-type eye morphogenesis
          erythrocyte development
          embryonic process involved in female pregnancy
          bone morphogenesis
          cardiac septum morphogenesis
          ventricular septum morphogenesis
          embryonic heart tube left/right pattern formation
          pulmonary artery morphogenesis
          cardiac neural crest cell development involved in heart development
          determination of heart left/right asymmetry
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          left/right axis specification
          nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          regulation of nucleic acid-templated transcription
          positive regulation of male gonad development
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          RNA polymerase II transcription corepressor activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription cofactor activity
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          histone acetyltransferase binding
          SMAD binding
          LBD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0