Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Resp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009049.1 | NP_033075.1 | 3 | 305 | ||
GenBank mRNA | AK012477.1 | - | 3 | 258 | ||
AK019440.1 | - | 3 | 255 | |||
BC027535.1 | - | 3 | 268 | 3376899 | ||
FJ621497.1 | - | 3 | 241 | |||
L34214.1 | - | 3 | 305 |