Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam193b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145382.4 | NP_663357.2 | 8 | 3612 | ||
GenBank mRNA | AK040568.1 | - | 2 | 1740 | ||
AK155237.1 | - | 8 | 3606 | |||
AK173303.1 | - | 8 | 2461 | |||
BC021381.1 | - | 2 | 1202 | 5043282 | ||
BC023015.1 | - | 2 | 1144 | 5364758 | ||
BC025483.1 | - | 8 | 2471 | 5150988 |